5-Bromo-2-hydroxypyrimidine 98%

Warning

Synonyms: 5-Bromopyrimidin-2(1H)-one5-Bromo-2-pyrimidinone

L18347-03 L18347-06 L18347-06 L18347-03
AAAL18347-06EA 214.23 USD
AAAL18347-06 CAAAAL18347-06 AAAL18347-03 CAAAAL18347-03
5-Bromo-2-hydroxypyrimidine 98%
5-Bromo-2-hydroxypyrimidine
Formula: C₄H₃BrN₂O
MW: 174.98 g/mol
Melting Pt: ca 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01318956
CAS Number: 38353-06-9
EINECS: 253-896-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR