3-Methylthiophene-2-carbonyl chloride 98%

Danger

L17989-04 L17989-09 L17989-04 L17989-09
AAAL17989-09EA 135.42 USD
CAAAAL17989-04 AAAL17989-09 CAAAAL17989-09 AAAL17989-04
3-Methylthiophene-2-carbonyl chloride 98%
3-Methylthiophene-2-carbonyl chloride
Formula: C₆H₅ClOS
Boiling Pt: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00130091
CAS Number: 61341-26-2
EINECS: 000-000-0
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR