1-Chloroisoquinoline 95%

Supplier: Thermo Scientific Chemicals

Warning

L17831-03 L17831-06 L17831-03 L17831-06
AAAL17831-03EA 42.3 USD
CAAAAL17831-03 AAAL17831-03 CAAAAL17831-06 AAAL17831-06
1-Chloroisoquinoline 95%
1-Chloroisoquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 105…108 °C/4 mm
Melting Pt: 33…35 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00024134
CAS Number: 19493-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR