8-Amino-1-naphthalenesulfonic acid 98%

Synonyms: 8-Aminonaphthalene-1-sulfonic acid8-Aminonaphthalene-1-sulphonic acid

L17616-36 L17616-14 L17616-22 L17616-14 L17616-22 L17616-36
AAAL17616-14EA 35.13 USD
CAAAAL17616-36 CAAAAL17616-14 AAAL17616-14 AAAL17616-36 CAAAAL17616-22 AAAL17616-22
8-Amino-1-naphthalenesulfonic acid 98%
8-Amino-1-naphthalenesulphonic acid
Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00035730
CAS Number: 82-75-7
EINECS: 201-437-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR