5-Bromo-7-nitroindoline 98%

Supplier: Thermo Scientific Chemicals
Warning

L16993-06 L16993-03 L16993-03 L16993-06
AAAL16993-03EA 53.69 USD
AAAL16993-03 CAAAAL16993-06 CAAAAL16993-03 AAAL16993-06
5-Bromo-7-nitroindoline 98%
5-Bromo-7-nitroindoline
Formula: C₈H₇BrN₂O₂
MW: 243.06 g/mol
Melting Pt: 128…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00005708
CAS Number: 80166-90-1
EINECS: 279-411-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR