Dimethoxy(methyl)silane 97%

Danger

L16197-14 L16197-06 L16197-14 L16197-06
AAAL16197-06EA 61.3 USD
AAAL16197-06 CAAAAL16197-06 AAAL16197-14 CAAAAL16197-14
Dimethoxy(methyl)silane 97%
Dimethoxy(methyl)silane
Formula: C₃H₁₀O₂Si
Boiling Pt: 61 °C
Melting Pt: –136 °C
Density: 0.861
Flash Pt: 2 °C(35 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048019
CAS Number: 16881-77-9
EINECS: 240-914-9
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR