5-Methoxy-1-tetralone 97%

Synonyms: 5-Methoxy-3,4-dihydronaphthalenone

L15129-03 L15129-06 L15129-03 L15129-06
AAAL15129-06EA 104.03 USD
AAAL15129-06 CAAAAL15129-06 AAAL15129-03 CAAAAL15129-03
5-Methoxy-1-tetralone 97%
5-Methoxy-1-tetralone
Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 160…162 °C/7 mm
Melting Pt: 86…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00001692
CAS Number: 33892-75-0
EINECS: 251-723-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR