Triphenylphosphonium bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Triphenylphosphine hydrobromide

L14290-14 L14290-14
AAAL14290-14EA 91.79 USD
AAAL14290-14 CAAAAL14290-14
Triphenylphosphonium bromide 97%
Triphenylphosphonium bromide
Formula: C₁₈H₁₆BrP
Melting Pt: ca 196 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035107
CAS Number: 6399-81-1
EINECS: 229-012-6
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR