Methyl-2-(trifluoromethyl)benzoate 97%

Warning

Synonyms: Methyl 2-(trifluoromethyl)benzoate

L14132-03 L14132-06 L14132-03 L14132-06
AAAL14132-06EA 158.73 USD
AAAL14132-06 CAAAAL14132-03 AAAL14132-03 CAAAAL14132-06
Methyl-2-(trifluoromethyl)benzoate 97%
Methyl 2-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 104…105 °C/25 mm
Density: 1.306
Storage Temperature: Ambient
MDL Number: MFCD00236050
CAS Number: 344-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR