3-Fluoro-5-(trifluoromethyl)benzamide 97%

Warning

L12854-03 L12854-06 L12854-06 L12854-03
AAAL12854-03EA 66.44 USD
CAAAAL12854-06 AAAL12854-03 CAAAAL12854-03 AAAL12854-06
3-Fluoro-5-(trifluoromethyl)benzamide 97%
3-Fluoro-5-(trifluoromethyl)benzamide
Formula: C₈H₅F₄NO
Melting Pt: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00061149
CAS Number: 207986-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR