5-Methylhydantoin 98%

L11992-03 L11992-06 L11992-03 L11992-06
AAAL11992-06EA 155.53 USD
AAAL11992-06 AAAL11992-03 CAAAAL11992-06 CAAAAL11992-03
5-Methylhydantoin 98%
5-Methylhydantoin
Formula: C₄H₆N₂O₂
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00022398
CAS Number: 616-03-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR