2-Phosphonopropionic acid 98+%

Danger

L11588-09 L11588-09 L11588-04 L11588-04
AAAL11588-09EA 214.23 USD
AAAL11588-09 CAAAAL11588-09 AAAL11588-04 CAAAAL11588-04
2-Phosphonopropionic acid 98+%
2-Phosphonopropionic acid
Formula: C₃H₇O₅P
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00041415
CAS Number: 5962-41-4
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR