6-Methoxy-2-tetralone tech. 90%, Technical Grade

Warning

Synonyms: 3,4-Dihydro-6-methoxynaphthalen-2(1H)-one6-Methoxy-3,4-dihydronaphthalen-2(1H)-one

L11007-03 L11007-03 L11007-MD L11007-MD
AAAL11007-03EA 125 USD
AAAL11007-03 CAAAAL11007-MD CAAAAL11007-03 AAAL11007-MD
6-Methoxy-2-tetralone tech. 90%, Technical Grade
6-Methoxy-2-tetralone
Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 114…116 °C/0.2 mm
Melting Pt: ca 35 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001729
CAS Number: 2472-22-2
EINECS: 219-592-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR