(2,4-Dichlorophenoxy)acetonitrile 97%

Warning

L10869-03 L10869-03
AAAL10869-03EA 44.24 USD
AAAL10869-03 CAAAAL10869-03
(2,4-Dichlorophenoxy)acetonitrile 97%
(2,4-Dichlorophenoxy)acetonitrile
Formula: C₈H₅Cl₂NO
Melting Pt: 46…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00019826
CAS Number: 3956-63-6
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR