2-Phenoxyphenylacetonitrile 98%

Warning

L10194-03 L10194-03 L10194-06 L10194-06
AAAL10194-06EA 246.42 USD
AAAL10194-06 CAAAAL10194-03 AAAL10194-03 CAAAAL10194-06
2-Phenoxyphenylacetonitrile 98%
2-Phenoxyphenylacetonitrile
Formula: C₁₄H₁₁NO
Boiling Pt: 145 °C/1 mm
Density: 1.110
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016386
CAS Number: 25562-98-5
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR