2-Fluoro-3-(trifluoromethyl)benzamide 97%

Warning

L10135-03 L10135-06 L10135-03 L10135-06
AAAL10135-06EA 175.38 USD
CAAAAL10135-06 AAAL10135-06 CAAAAL10135-03 AAAL10135-03
2-Fluoro-3-(trifluoromethyl)benzamide 97%
2-Fluoro-3-(trifluoromethyl)benzamide
Formula: C₈H₅F₄NO
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00061146
CAS Number: 207853-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR