4-Benzyloxyphenylacetonitrile 97%

Warning

Synonyms: 2-(4-(Benzyloxy)phenyl)acetonitrile

L09771-03 L09771-03 L09771-MD L09771-MD L09771-06 L09771-06
AAAL09771-MDEA 34.23 USD
AAAL09771-MD AAAL09771-06 CAAAAL09771-MD CAAAAL09771-06 AAAL09771-03 CAAAAL09771-03
4-Benzyloxyphenylacetonitrile 97%
4-Benzyloxyphenylacetonitrile
Formula: C₁₅H₁₃NO
MW: 223.28 g/mol
Boiling Pt: 170…173 °C/0.1 mm
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00016400
CAS Number: 838-96-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR