4-Pentylbenzaldehyde 95%

Supplier: Thermo Scientific Chemicals

L09208-03 L09208-06 L09208-03 L09208-06
AAAL09208-06EA 181 USD
AAAL09208-06 CAAAAL09208-06 AAAL09208-03 CAAAAL09208-03
4-Pentylbenzaldehyde 95%
4-Pentylbenzaldehyde
Formula: C₁₂H₁₆O
Boiling Pt: 142…144 °C/2 mm
Storage Temperature: Ambient
MDL Number: MFCD00043599
CAS Number: 6853-57-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR