2,4,6-Tribromo-m-cresol 98+%

Warning

Synonyms: 2,4,6-Tribromo-3-methylphenol

L09183-14 L09183-22 L09183-22 L09183-14
AAAL09183-14EA 57.56 USD
AAAL09183-14 CAAAAL09183-22 AAAL09183-22 CAAAAL09183-14
2,4,6-Tribromo-m-cresol 98+%
2,4,6-Tribromo-m-cresol
Formula: C₇H₅Br₃O
MW: 344.84 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00019976
CAS Number: 4619-74-3
EINECS: 225-032-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR