Chlorthaldimethyl 97%

Supplier: Alfa Aesar
Warning

Synonyms: Dimethyl tetrachloroterephthalate

L08800-14 L08800-06 L08800-14 L08800-06
AAAL08800-14EA 156.48 USD
CAAAAL08800-14 CAAAAL08800-06 AAAL08800-14 AAAL08800-06
Chlorthaldimethyl 97%
Chlorthal-dimethyl
Formula: C₁₀H₆Cl₄O₄
Melting Pt: 156-159°C
Storage Temperature: Ambient
MDL Number: MFCD00014902
CAS Number: 1861-32-1
EINECS: 217-464-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR