2-Acetamido-6-chlorobenzoic acid 99%

Warning

L08632-03 L08632-06 L08632-06 L08632-14 L08632-14 L08632-03
AAAL08632-06EA 65.15 USD
AAAL08632-06 CAAAAL08632-14 CAAAAL08632-03 AAAL08632-03 AAAL08632-14 CAAAAL08632-06
2-Acetamido-6-chlorobenzoic acid 99%
2-Acetamido-6-chlorobenzoic acid
Formula: C₉H₈ClNO₃
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00051939
CAS Number: 19407-42-2
EINECS: 243-040-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR