Phthalamide 97%

Supplier: Thermo Scientific Chemicals

L08600-18 L08600-18 L08600-09 L08600-09
AAAL08600-18EA 119 USD
CAAAAL08600-18 AAAL08600-18 CAAAAL08600-09 AAAL08600-09
Phthalamide 97%
Phthalamide
Formula: C₈H₈N₂O₂
Melting Pt: 223 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00025478
CAS Number: 88-96-0
EINECS: 201-870-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR