2-(Allylthio)nicotinic acid 98%

Supplier: Thermo Scientific Chemicals

Warning

L07090-06 L07090-06
AAAL07090-06EA 48.9 USD
CAAAAL07090-06 AAAL07090-06
2-(Allylthio)nicotinic acid 98%
2-(Allylthio)nicotinic acid
Formula: C₉H₉NO₂S
Melting Pt: 144…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00051681
CAS Number: 175135-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR