2-Acetyl-5-nitrobenzofuran 98%

L06308-03 L06308-03 L06308-06 L06308-06
AAAL06308-03EA 79.74 USD
CAAAAL06308-06 CAAAAL06308-03 AAAL06308-03 AAAL06308-06
2-Acetyl-5-nitrobenzofuran 98%
2-Acetyl-5-nitrobenzofuran
Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
Melting Pt: 173…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00051634
CAS Number: 23136-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR