2-Naphthonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Naphthalene-2-carbonitrile

L03704-14 L03704-14
AAAL03704-14EA 227.99 USD
CAAAAL03704-14 AAAL03704-14
2-Naphthonitrile 97%
2-Naphthonitrile
Formula: C₁₁H₇N
MW: 153.18 g/mol
Boiling Pt: 156…158 °C/12 mm
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00016807
CAS Number: 613-46-7
EINECS: 210-344-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR