(2,5-Dimethylphenyl)acetonitrile 98+%

Warning

L01942-06 L01942-14 L01942-14 L01942-06
AAAL01942-14EA 188.81 USD
AAAL01942-14 CAAAAL01942-06 CAAAAL01942-14 AAAL01942-06
(2,5-Dimethylphenyl)acetonitrile 98+%
(2,5-Dimethylphenyl)acetonitrile
Formula: C₁₀H₁₁N
Boiling Pt: 73 °C/1 mm
Melting Pt: 27…29 °C
Storage Temperature: Ambient
MDL Number: MFCD00013816
CAS Number: 16213-85-7
EINECS: 240-339-3
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR