4-(Diethylamino)benzonitrile 97%

Warning

L01905-14 L01905-06 L01905-14 L01905-06
AAAL01905-14EA 166.5 USD
AAAL01905-14 CAAAAL01905-06 CAAAAL01905-14 AAAL01905-06
4-(Diethylamino)benzonitrile 97%
4-(Diethylamino)benzonitrile
Formula: C₁₁H₁₄N₂
Melting Pt: 66…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00019754
CAS Number: 2873-90-7
EINECS: 220-712-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR