(4-Methylbenzyl)triphenylphosphonium bromide 98+%

Warning

L01562-14 L01562-06 L01562-14 L01562-06
AAAL01562-06EA 42.22 USD
AAAL01562-06 CAAAAL01562-06 AAAL01562-14 CAAAAL01562-14
(4-Methylbenzyl)triphenylphosphonium bromide 98+%
(4-Methylbenzyl)triphenylphosphonium bromide
Formula: C₂₆H₂₄BrP
Melting Pt: 268…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00031580
CAS Number: 2378-86-1
EINECS: 219-159-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR