(2-Hydroxybenzyl)triphenylphosphonium bromide 98+%

Warning

L00778-09 L00778-18 L00778-18 L00778-09
AAAL00778-18EA 263.81 USD
CAAAAL00778-09 AAAL00778-18 CAAAAL00778-18 AAAL00778-09
(2-Hydroxybenzyl)triphenylphosphonium bromide 98+%
(2-Hydroxybenzyl)triphenylphosphonium bromide
Formula: C₂₅H₂₂BrOP
Melting Pt: 240…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00011902
CAS Number: 70340-04-4
EINECS: 274-574-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR