4-Bromo-N-phenylbenzamide 98%

Warning

H50597-03 H50597-MD H50597-03 H50597-MD
AAAH50597-03EA 240.43 USD
CAAAAH50597-03 AAAH50597-03 CAAAAH50597-MD AAAH50597-MD
4-Bromo-N-phenylbenzamide 98%
4-Bromo-N-phenylbenzamide
Formula: C₁₃H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD00624477
CAS Number: 6846-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR