2-Acetyl-5-bromobenzofuran 99%

Warning

Synonyms: 1-(5-Bromobenzofuran-2-yl)ethanone2-Acetyl-5-bromobenzo[b]furan

H50593-06 H50593-03 H50593-03 H50593-06
AAAH50593-06EA 357.42 USD
AAAH50593-06 CAAAAH50593-06 AAAH50593-03 CAAAAH50593-03
2-Acetyl-5-bromobenzofuran 99%
2-Acetyl-5-bromobenzofuran
Formula: C₁₀H₇BrO₂
MW: 239.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00276737
CAS Number: 38220-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR