2-Bromo-6-ethoxynaphthalene 98%

Warning

H50568-03 H50568-MD H50568-MD H50568-03
AAAH50568-MDEA 78.24 USD
AAAH50568-MD CAAAAH50568-MD AAAH50568-03 CAAAAH50568-03
2-Bromo-6-ethoxynaphthalene 98%
2-Bromo-6-ethoxynaphthalene
Formula: C₁₂H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD01568865
CAS Number: 66217-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR