2-Chloro-3-methylquinoline 97%

Warning

H50492-03 H50492-03 H50492-06 H50492-06
AAAH50492-03EA 96.16 USD
CAAAAH50492-03 AAAH50492-03 CAAAAH50492-06 AAAH50492-06
2-Chloro-3-methylquinoline 97%
2-Chloro-3-methylquinoline
Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00100265
CAS Number: 57876-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR