2-Chloro-6-methoxyisonicotinic acid 96%

Warning

Synonyms: 2-Chloro-6-methoxypyridine-4-carboxylic acid

H50168-06 H50168-03 H50168-03 H50168-06
AAAH50168-03EA 66.12 USD
AAAH50168-03 CAAAAH50168-03 AAAH50168-06 CAAAAH50168-06
2-Chloro-6-methoxyisonicotinic acid 96%
2-Chloro-6-methoxyisonicotinic acid
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Boiling Pt: 209…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00173928
CAS Number: 15855-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR