3-Amino-4-formylpyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Aminopyridine-4-carboxaldehyde3-Aminoisonicotinaldehyde3-Amino-pyridine-4-carbaldehyde

H50024-03 H50024-03 H50024-MD H50024-MD
AAAH50024-MDEA 87.37 USD
CAAAAH50024-03 AAAH50024-MD AAAH50024-03 CAAAAH50024-MD
3-Amino-4-formylpyridine 95%
3-Amino-4-formylpyridine
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Melting Pt: ca 92 °C
Storage Temperature: Ambient
MDL Number: MFCD06410684
CAS Number: 55279-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR