3-Chloro-5-methoxyphenylboronic acid 94%

Warning

Synonyms: 3-Chloro-5-methoxybenzeneboronic acid

H27442-MD H27442-03
AAH27442-03EA 269.72 USD
AAH27442-03 AAH27442-MD
3-Chloro-5-methoxyphenylboronic acid 94%
3-Chloro-5-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD03095046
CAS Number: 915201-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR