2-Chloro-5-propylpyrimidine 98%

Warning

H27409-09 H27409-03
AAH27409-03EA 49.45 USD
AAH27409-03 AAH27409-09
2-Chloro-5-propylpyrimidine 98%
2-Chloro-5-propylpyrimidine
Formula: C₇H₉ClN₂
Boiling Pt: 100 °C/3 mm
Density: 1.142
Storage Temperature: Ambient
MDL Number: MFCD00799403
CAS Number: 219555-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR