Tetrabutylammonium dihydrogentrifluoride tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals

Danger

H27266-06
AAH27266-06EA 150 USD
AAH27266-06
Tetrabutylammonium dihydrogentrifluoride tech. 90%, Technical Grade
Tetrabutylammonium dihydrogentrifluoride
Formula: C₁₆H₃₈F₃N
Storage Temperature: Ambient
MDL Number: MFCD00145365
CAS Number: 99337-56-1
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR