2-Chloro-6-hydrazinopyridine 95%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: (6-Chloro-2-pyridyl)hydrazine

H27249-MD H27249-03
AAH27249-03EA 211 USD
AAH27249-03 AAH27249-MD
2-Chloro-6-hydrazinopyridine 95%
2-Chloro-6-hydrazinopyridine
Formula: C₅H₆ClN₃
Boiling Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00214724
CAS Number: 5193-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR