6,2'-Dibromo-3,4'-bipyridyl 95%

Warning

Synonyms: 2',6-Dibromo-3,4'-bipyridine

H27214-MC H27214-ME
AAH27214-MEEA 407.37 USD
AAH27214-ME AAH27214-MC
6,2'-Dibromo-3,4'-bipyridyl 95%
6,2'-Dibromo-3,4'-bipyridyl
Formula: C₁₀H₆Br₂N₂
Melting Pt: 189…193 °C
Storage Temperature: Ambient
MDL Number: MFCD09037424
CAS Number: 942206-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR