2,5-Dichloro-4-pyridineboronic acid 95%

Warning

Synonyms: 2,5-Dichloropyridine-4-boronic acid2,5-Dichloro-4-pyridylboronic acid

H27170-03
AAH27170-03EA 428.04 USD
AAH27170-03
2,5-Dichloro-4-pyridineboronic acid 95%
2,5-Dichloro-4-pyridineboronic acid
Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
Melting Pt: ca 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06798254
CAS Number: 847664-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR