2,6-Dichloro-4-nitropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H27131-03 H27131-06
AAH27131-03EA 38.01 USD
AAH27131-03 AAH27131-06
2,6-Dichloro-4-nitropyridine 97%
2,6-Dichloro-4-nitropyridine
Formula: C₅H₂Cl₂N₂O₂
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD05670545
CAS Number: 25194-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR