5-Chloro-2-methoxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-2-methoxybenzeneboronic acid(5-Chloro-2-methoxyphenyl)boronic acid

H27127-06 H27127-14
AAH27127-06EA 71.79 USD
AAH27127-06 AAH27127-14
5-Chloro-2-methoxyphenylboronic acid 97%
5-Chloro-2-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Melting Pt: 134…141 °C
Storage Temperature: Ambient
MDL Number: MFCD01318966
CAS Number: 89694-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR