Isopropylphosphonic dichloride tech. 90%, Technical Grade

Danger

H27109-03 H27109-06
AAH27109-06EA 224.44 USD
AAH27109-06 AAH27109-03
Isopropylphosphonic dichloride tech. 90%, Technical Grade
Isopropylphosphonic dichloride
Formula: C₃H₇Cl₂OP
Storage Temperature: Ambient
MDL Number: MFCD00603000
CAS Number: 1498-46-0
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR