2-Amino-5-iodo-4-methylpyridine 95%

Warning

Synonyms: 5-Iodo-4-methylpyridin-2-ylamine5-Iodo-4-methyl-pyridin-2-ylamine

H27089-03 H27089-MD
AAH27089-MDEA 89.63 USD
AAH27089-MD AAH27089-03
2-Amino-5-iodo-4-methylpyridine 95%
2-Amino-5-iodo-4-methylpyridine
Formula: C₆H₇IN₂
MW: 234.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06637727
CAS Number: 356561-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR