3-Bromoisonicotinonitrile 95%

Danger

H27049-03 H27049-MD
AAH27049-MDEA 213.45 USD
AAH27049-MD AAH27049-03
3-Bromoisonicotinonitrile 95%
3-Bromoisonicotinonitrile
Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00234143
CAS Number: 13958-98-0
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR