N,N-Diethylaminophosphoryl dichloride 96%

Danger

Synonyms: Diethylphosphoramidic dichloride

H26972-03
AAH26972-03EA 32.19 USD
AAH26972-03
N,N-Diethylaminophosphoryl dichloride 96%
N,N-Diethylaminophosphoryl dichloride
Formula: C₄H₁₀Cl₂NOP
Boiling Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00013897
CAS Number: 1498-54-0
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR