5-Chloro-2-fluoropyridine 97%

Danger

H26945-06 H26945-14
AAH26945-14EA 198.79 USD
AAH26945-14 AAH26945-06
5-Chloro-2-fluoropyridine 97%
5-Chloro-2-chloropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
Boiling Pt: 149 °C
Density: 1.311
Flash Pt: 52 °C(126 °F)
Storage Temperature: Ambient
MDL Number: MFCD04112513
CAS Number: 1480-65-5
UN: 1992
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR