2-Chloro-4-(trifluoromethyl)benzonitrile 98%

Danger

H26894-14 H26894-03 H26894-06
AAH26894-06EA 105.89 USD
AAH26894-06 AAH26894-14 AAH26894-03
2-Chloro-4-(trifluoromethyl)benzonitrile 98%
2-Chloro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
MW: 205.57 g/mol
Boiling Pt: 192…193 °C
Melting Pt: 82…83 °C/8mm
Density: 1.389
Flash Pt: >104 °C(220 °F)
Storage Temperature: Ambient
MDL Number: MFCD00084939
CAS Number: 1813-33-8
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR