2,4-Bis(trifluoromethyl)benzonitrile 97%

Danger

H26869-03
AAH26869-03EA 132.09 USD
AAH26869-03
2,4-Bis(trifluoromethyl)benzonitrile 97%
2,4-Bis(trifluoromethyl)benzonitrile
Formula: C₉H₃F₆N
MW: 239.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061225
CAS Number: 177952-38-4
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR